Product Details
- SNP ID
-
rs77406959
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:105279907 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTTAAATAGGACTTGTTTCCTTA[A/C]GTGCAAATTTTCTCTAAGTTAGATA
- Phenotype
-
MIM: 600400
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PREP
PubMed Links
Gene Details
- Gene
- PREP
- Gene Name
- prolyl endopeptidase
View Full Product Details