Product Details

SNP ID
rs75343336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20688217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTAGAAGCTGAGAAGTTGAGTGT[A/C]GGGGGAGGGGCAGTGTACTAATAGC
Phenotype
MIM: 105850 MIM: 601030
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANG PubMed Links

Gene Details

Gene
ANG
Gene Name
angiogenin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097577.2 Intron NP_001091046.1
NM_001145.4 Intron NP_001136.1
Gene
RNASE4
Gene Name
ribonuclease A family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282192.1 Intron NP_001269121.1
NM_001282193.1 Intron NP_001269122.1
NM_002937.4 Intron NP_002928.1
NM_194431.2 Intron NP_919412.1

View Full Product Details