Product Details

SNP ID
rs75381963
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52430879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAGGGTGCTGAGAGTAGCAAC[A/G]GGTCTATGTTATAAGGAAATATATG
Phenotype
MIM: 616179
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TXNDC16 PubMed Links

Gene Details

Gene
TXNDC16
Gene Name
thioredoxin domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160047.1 4337 UTR 3 NP_001153519.1
NM_020784.2 4337 UTR 3 NP_065835.2
XM_017021505.1 4337 Intron XP_016876994.1

View Full Product Details