Product Details

SNP ID
rs75774651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55367957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCAACTACAATTATTATGATGA[A/G]AAAAGAAGCTGGGGCATGGCAAACC
Phenotype
MIM: 613515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG14 PubMed Links

Gene Details

Gene
ATG14
Gene Name
autophagy related 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014924.4 2881 UTR 3 NP_055739.2
XM_011536563.2 2881 UTR 3 XP_011534865.1

View Full Product Details