Product Details
- SNP ID
-
rs75225332
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55767578 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGAGTTTTATATTAGTCCAGAGG[G/T]TATTATATTAGCCCAGGCAAGAACT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPL13AP3
PubMed Links
Gene Details
- Gene
- RPL13AP3
- Gene Name
- ribosomal protein L13a pseudogene 3
There are no transcripts associated with this gene.
View Full Product Details