Product Details

SNP ID
rs78494962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64746842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGGTGTGGACTGGAGCTGTGT[C/T]AACCTCGTTTGGGAATACTGTCAAA
Phenotype
MIM: 182870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG3 PubMed Links

Gene Details

Gene
PLEKHG3
Gene Name
pleckstrin homology and RhoGEF domain containing G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308147.1 9720 Intron NP_001295076.1
Gene
SPTB
Gene Name
spectrin beta, erythrocytic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000347.5 9720 Intron NP_000338.3
NM_001024858.2 9720 UTR 3 NP_001020029.1
XM_005268023.4 9720 UTR 3 XP_005268080.1
XM_011537105.2 9720 Intron XP_011535407.1
XM_017021612.1 9720 UTR 3 XP_016877101.1
XM_017021613.1 9720 Intron XP_016877102.1
XM_017021614.1 9720 Intron XP_016877103.1

View Full Product Details