Product Details

SNP ID
rs78204860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100954384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGTCCTAAGCCTGAGCCAGTG[A/G]TGAAAACTGGTGGCATAGAAGTCAA
Phenotype
MIM: 613651
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORD113-9 PubMed Links

Gene Details

Gene
SNORD113-9
Gene Name
small nucleolar RNA, C/D box 113-9
There are no transcripts associated with this gene.

Gene
SNORD114-1
Gene Name
small nucleolar RNA, C/D box 114-1
There are no transcripts associated with this gene.

Gene
SNORD114-2
Gene Name
small nucleolar RNA, C/D box 114-2
There are no transcripts associated with this gene.

Gene
SNORD114-3
Gene Name
small nucleolar RNA, C/D box 114-3
There are no transcripts associated with this gene.

Gene
SNORD114-4
Gene Name
small nucleolar RNA, C/D box 114-4
There are no transcripts associated with this gene.

Gene
SNORD114-5
Gene Name
small nucleolar RNA, C/D box 114-5
There are no transcripts associated with this gene.

Gene
SNORD114-6
Gene Name
small nucleolar RNA, C/D box 114-6
There are no transcripts associated with this gene.

Gene
SNORD114-7
Gene Name
small nucleolar RNA, C/D box 114-7
There are no transcripts associated with this gene.

View Full Product Details