Product Details
- SNP ID
-
rs78396662
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:102935814 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAATTATGAGGAATGTAAATAACG[A/G]CAACCCATAGAACAAGAGAAGACAT
- Phenotype
-
MIM: 605799
MIM: 614062
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs552509293] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMN
- Gene Name
- amnion associated transmembrane protein
There are no transcripts associated with this gene.
- Gene
- CDC42BPB
- Gene Name
- CDC42 binding protein kinase beta
View Full Product Details