Product Details

SNP ID
rs78534977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103336181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGAGTCTTCAAAGTCTTTGAGC[C/T]GCAAAACTTGTTCTAATTGTATGCT
Phenotype
MIM: 601710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF5 PubMed Links

Gene Details

Gene
EIF5
Gene Name
eukaryotic translation initiation factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001969.4 Intron NP_001960.2
NM_183004.4 Intron NP_892116.2
Gene
LOC105370687
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
SNORA28
Gene Name
small nucleolar RNA, H/ACA box 28
There are no transcripts associated with this gene.

View Full Product Details