Product Details

SNP ID
rs75570865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113459265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGGCAGGAGAAGTTAACGTTCT[C/T]AGGTGTAAGCATTTCGGTCAATCAT
Phenotype
MIM: 610620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADPRHL1 PubMed Links

Gene Details

Gene
ADPRHL1
Gene Name
ADP-ribosylhydrolase like 1
There are no transcripts associated with this gene.

Gene
DCUN1D2
Gene Name
defective in cullin neddylation 1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014283.1 Intron NP_001014305.1
XM_017020643.1 Intron XP_016876132.1
XM_017020644.1 Intron XP_016876133.1
XM_017020645.1 Intron XP_016876134.1
XM_017020646.1 Intron XP_016876135.1
XM_017020647.1 Intron XP_016876136.1
XM_017020648.1 Intron XP_016876137.1
XM_017020649.1 Intron XP_016876138.1

View Full Product Details