Product Details

SNP ID
rs75194939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52690483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGAGGCAGGAATAGGCCGCTG[G/T]TGCAGAGGTTTTCCTTACCAAGCTA
Phenotype
MIM: 611158
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRT77 PubMed Links

Gene Details

Gene
KRT77
Gene Name
keratin 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175078.2 2210 UTR 3 NP_778253.2
XM_011538288.2 2210 UTR 3 XP_011536590.1
XM_011538289.2 2210 Intron XP_011536591.1

View Full Product Details