Product Details

SNP ID
rs77287121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55426493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTGTAATCCTAGATTTCTGAT[C/G]AGCATTCTAACAGGGGACAAATCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR6C76 PubMed Links

Gene Details

Gene
OR6C76
Gene Name
olfactory receptor family 6 subfamily C member 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005183.1 240 Missense Mutation ATC,ATG I80M NP_001005183.1

View Full Product Details