Product Details

SNP ID
rs78062104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55977889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTGTCTCCTGGTTCTGTGGTG[G/T]TATGGCTCCTTTTTTTAAACTTGTG
Phenotype
MIM: 116953 MIM: 179514
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK2 PubMed Links

Gene Details

Gene
CDK2
Gene Name
cyclin dependent kinase 2
There are no transcripts associated with this gene.

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252036.1 Intron NP_001238965.1
NM_001252037.1 Intron NP_001238966.1
NM_002868.3 Intron NP_002859.1
XM_005269051.2 Intron XP_005269108.1
XM_005269052.2 Intron XP_005269109.1

View Full Product Details