Product Details

SNP ID
rs74609971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76052276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAACAAACAAACAAAAAAAAA[C/T]CCCAAACTTGAACACAAACTCCAAC
Phenotype
MIM: 164060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAP1L1 PubMed Links

Gene Details

Gene
NAP1L1
Gene Name
nucleosome assembly protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719425.3 Intron XP_006719488.1
XM_011538390.2 Intron XP_011536692.2
XM_011538393.2 Intron XP_011536695.1
XM_017019338.1 Intron XP_016874827.1
XM_017019339.1 Intron XP_016874828.1
XM_017019340.1 Intron XP_016874829.1

View Full Product Details