Product Details

SNP ID
rs74956555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:94971468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTAAGGTGTTGAAGGTGGGATC[C/T]ACTCCTGAATCTTCTTTCTAGTGGT
Phenotype
MIM: 614530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA12 PubMed Links

Gene Details

Gene
NDUFA12
Gene Name
NADH:ubiquinone oxidoreductase subunit A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258338.1 355 UTR 3 NP_001245267.1
NM_018838.4 355 Nonsense Mutation TAG,TGG *137W NP_061326.1

View Full Product Details