Product Details

SNP ID
rs75679077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101400660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGCACTGTAAACACTGGGAAG[A/G]ATAAAAAACGATAAACGTGAGGCCA
Phenotype
MIM: 603425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL1 PubMed Links

Gene Details

Gene
ARL1
Gene Name
ADP ribosylation factor like GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177.5 Intron NP_001168.1
NM_001301068.1 Intron NP_001287997.1

View Full Product Details