Product Details

SNP ID
rs78074890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:50807146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATACTTAAGAGAATTAAAGATG[C/T]TTGGGATAGGGTCCAAGAAAGAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A1CF PubMed Links

Gene Details

Gene
A1CF
Gene Name
APOBEC1 complementation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198818.1 Intron NP_001185747.1
NM_001198819.1 Intron NP_001185748.1
NM_001198820.1 Intron NP_001185749.1
NM_014576.3 Intron NP_055391.2
NM_138932.2 Intron NP_620310.1
NM_138933.2 Intron NP_620311.1
XM_005269718.2 Intron XP_005269775.1
XM_005269720.3 Intron XP_005269777.1
XM_011539729.2 Intron XP_011538031.1
XM_011539730.2 Intron XP_011538032.1
XM_017016160.1 Intron XP_016871649.1

View Full Product Details