Product Details

SNP ID
rs77913769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89432902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCTAGTCCAGCTTTCCTTATT[A/T]TGTGCTGTTTTCCCTTATAAATATT
Phenotype
MIM: 611910
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC16A12 PubMed Links

Gene Details

Gene
SLC16A12
Gene Name
solute carrier family 16 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213606.3 1902 UTR 3 NP_998771.3
XM_017016237.1 1902 UTR 3 XP_016871726.1
XM_017016238.1 1902 UTR 3 XP_016871727.1
XM_017016239.1 1902 UTR 3 XP_016871728.1

View Full Product Details