Product Details

SNP ID
rs75632981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94546657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTTTTTGTATGTTTACAGATA[C/T]TGCCTTTGAGAATCCTTTCGCAAGT
Phenotype
MIM: 603946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HELLS PubMed Links

Gene Details

Gene
HELLS
Gene Name
helicase, lymphoid-specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289067.1 Intron NP_001275996.1
NM_001289068.1 Intron NP_001275997.1
NM_001289069.1 Intron NP_001275998.1
NM_001289070.1 Intron NP_001275999.1
NM_001289071.1 Intron NP_001276000.1
NM_001289072.1 Intron NP_001276001.1
NM_001289073.1 Intron NP_001276002.1
NM_001289074.1 Intron NP_001276003.1
NM_001289075.1 Intron NP_001276004.1
NM_018063.4 Intron NP_060533.2

View Full Product Details