Product Details

SNP ID
rs76168984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123137362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAAGACGCGCACAGTCTTCTC[C/G]CGCAGCCAGGTCTTCCAGCTCGAGT
Phenotype
MIM: 600647 MIM: 613380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMX2 PubMed Links

Gene Details

Gene
HMX2
Gene Name
H6 family homeobox 2
There are no transcripts associated with this gene.

Gene
HMX3
Gene Name
H6 family homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105574.1 705 Silent Mutation TCC,TCG S235S NP_001099044.1

View Full Product Details