Product Details

SNP ID
rs74532659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123137518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACAAGTGGAAGCGGCAGCTGGC[A/G]GCGGAGCTGGAGGCGGCCAACCTGA
Phenotype
MIM: 600647 MIM: 613380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMX2 PubMed Links

Gene Details

Gene
HMX2
Gene Name
H6 family homeobox 2
There are no transcripts associated with this gene.

Gene
HMX3
Gene Name
H6 family homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105574.1 861 Silent Mutation GCA,GCG A287A NP_001099044.1

View Full Product Details