Product Details
- SNP ID
-
rs79038092
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:45003700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTGTGGCAGGAAGAGGGTGCTGG[A/G]GCAAGTGGAAGACTCGGGCAGCGCG
- Phenotype
-
MIM: 613521
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HECTD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs562126758] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HECTD3
- Gene Name
- HECT domain E3 ubiquitin protein ligase 3
- Gene
- UROD
- Gene Name
- uroporphyrinogen decarboxylase
There are no transcripts associated with this gene.
View Full Product Details