Product Details

SNP ID
rs78920799
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:45016450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCCATGATTTATTCATAGAAAA[C/G]CACTGTGAATTCACACACTTGCTAA
Phenotype
MIM: 613521
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HECTD3 PubMed Links

Gene Details

Gene
HECTD3
Gene Name
HECT domain E3 ubiquitin protein ligase 3
There are no transcripts associated with this gene.

Gene
UROD
Gene Name
uroporphyrinogen decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000374.4 5790 Intron NP_000365.3
XM_005271169.1 5790 Intron XP_005271226.1
XM_005271170.1 5790 Intron XP_005271227.1
Gene
ZSWIM5
Gene Name
zinc finger SWIM-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020883.1 5790 UTR 3 NP_065934.1
XM_011541861.2 5790 Intron XP_011540163.1
XM_017001913.1 5790 Intron XP_016857402.1

View Full Product Details