Product Details

SNP ID
rs77163359
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:78628271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTGGTGTGTCGTTTATCGAAAA[C/T]GGATATTTTCATTATATGTCGAGAT
Phenotype
MIM: 613975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI44L PubMed Links

Gene Details

Gene
IFI44L
Gene Name
interferon induced protein 44 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006820.3 394 Missense Mutation ACG,ATG T119M NP_006811.2
XM_005270391.3 394 Intron XP_005270448.1
XM_005270392.4 394 Intron XP_005270449.1
XM_005270393.4 394 Intron XP_005270450.1
XM_006710304.3 394 Intron XP_006710367.1
XM_011540539.2 394 Missense Mutation ACG,ATG T119M XP_011538841.1
XM_017000120.1 394 Intron XP_016855609.1

View Full Product Details