Product Details

SNP ID
rs78129995
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:87332065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCGCCCCCGGTGACGGCCGGCT[C/G]CCTCTCCTGGAAGCGGTGCGCAGGC
Phenotype
MIM: 603129
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LMO4 PubMed Links

Gene Details

Gene
LMO4
Gene Name
LIM domain only 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006769.3 3000 Missense Mutation TCC,TGC S17C NP_006760.1
XM_005271291.3 3000 Missense Mutation TCC,TGC S17C XP_005271348.1

View Full Product Details