Product Details

SNP ID
rs79501187
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:93889080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGACCGAATACCGCAGTAGCCA[C/G]AGCGGCACCCACTCGTGCGCTTGAA
Phenotype
MIM: 611199 MIM: 601176
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
There are no transcripts associated with this gene.

Gene
GCLM
Gene Name
glutamate-cysteine ligase modifier subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308253.1 1104 Silent Mutation CTC,CTG L223L NP_001295182.1
NM_002061.3 1104 Silent Mutation CTC,CTG L245L NP_002052.1
XM_011541261.2 1104 Silent Mutation CTC,CTG L157L XP_011539563.1
XM_017001056.1 1104 UTR 3 XP_016856545.1
XM_017001057.1 1104 UTR 3 XP_016856546.1

View Full Product Details