Product Details

SNP ID
rs79009722
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:36796266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGAGCCAGGGACAGAGATCTG[C/T]AGCCTCCTGGGGAGGGGGCACTCTG
Phenotype
MIM: 138243
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GRIK3 PubMed Links

Gene Details

Gene
GRIK3
Gene Name
glutamate ionotropic receptor kainate type subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000831.3 8480 UTR 3 NP_000822.2

View Full Product Details