Product Details

SNP ID
rs76711372
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:108570864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGAACAGTTGGAGGATGAAGGC[C/T]TTAGTGATGTTAAAGGAAGATATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM102B PubMed Links

Gene Details

Gene
FAM102B
Gene Name
family with sequence similarity 102 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010883.2 Intron NP_001010883.2

View Full Product Details