Product Details

SNP ID
rs79471201
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:109494214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGACCCGCTGGCTGCGGAGAGG[C/T]AGTGGGATCTTGGCGCACCTGGTAG
Phenotype
MIM: 615689
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMIGO1 PubMed Links

Gene Details

Gene
AMIGO1
Gene Name
adhesion molecule with Ig like domain 1
There are no transcripts associated with this gene.

Gene
ATXN7L2
Gene Name
ataxin 7 like 2
There are no transcripts associated with this gene.

Gene
CYB561D1
Gene Name
cytochrome b561 family member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134400.1 136 Silent Mutation GGC,GGT G25G NP_001127872.1
NM_001134402.1 136 Intron NP_001127874.1
NM_001134403.1 136 Silent Mutation GGC,GGT G25G NP_001127875.1
NM_001134404.1 136 Silent Mutation GGC,GGT G25G NP_001127876.1
NM_182580.2 136 Silent Mutation GGC,GGT G25G NP_872386.1
XM_005270775.2 136 UTR 5 XP_005270832.1
XM_005270776.3 136 Intron XP_005270833.1
XM_005270777.2 136 UTR 5 XP_005270834.1
XM_011541286.2 136 Intron XP_011539588.1
XM_011541287.2 136 Intron XP_011539589.2
XM_017001078.1 136 Intron XP_016856567.1
XM_017001079.1 136 Intron XP_016856568.1

View Full Product Details