Product Details

SNP ID
rs79760013
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:37611887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACTCTGGCTCACCAGGTCTGGA[A/G]TGAGACCTGAGATTCTCATTCCTAA
Phenotype
MIM: 609595
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSPO1 PubMed Links

Gene Details

Gene
RSPO1
Gene Name
R-spondin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038633.3 2552 UTR 3 NP_001033722.1
NM_001242908.1 2552 UTR 3 NP_001229837.1
NM_001242909.1 2552 UTR 3 NP_001229838.1
NM_001242910.1 2552 UTR 3 NP_001229839.1
XM_006710583.3 2552 UTR 3 XP_006710646.1

View Full Product Details