Product Details

SNP ID
rs77223842
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:119795331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGAGGCAGGATGCAGTGAGGA[C/T]CTGAGTGTGGCCTGTCCTCAAGCCC
Phenotype
MIM: 609846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REG4 PubMed Links

Gene Details

Gene
REG4
Gene Name
regenerating family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159352.1 Intron NP_001152824.1
NM_001159353.1 Intron NP_001152825.1
NM_032044.3 Intron NP_114433.1

View Full Product Details