Product Details

SNP ID
rs79002669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111545271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTATCAGTGCAGCTGGTCCCA[A/G]GGCCACAGATGGTACTTATTGACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf88 PubMed Links

Gene Details

Gene
C11orf88
Gene Name
chromosome 11 open reading frame 88
There are no transcripts associated with this gene.

Gene
LAYN
Gene Name
layilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258390.1 Intron NP_001245319.1
NM_001258391.1 Intron NP_001245320.1
NM_001318799.1 Intron NP_001305728.1
NM_178834.4 Intron NP_849156.1
XM_006718769.3 Intron XP_006718832.1
XM_006718770.3 Intron XP_006718833.1

View Full Product Details