Product Details

SNP ID
rs77825215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122097031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAATTAATACCAATCAAACATT[C/T]TCTTTCCCTGAGGCTTGATGTGTTC
Phenotype
MIM: 615965 MIM: 610104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR100HG PubMed Links

Gene Details

Gene
MIR100HG
Gene Name
mir-100-let-7a-2 cluster host gene
There are no transcripts associated with this gene.

Gene
MIR125B1
Gene Name
microRNA 125b-1
There are no transcripts associated with this gene.

View Full Product Details