Product Details

SNP ID
rs76401585
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123631943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAAGCCCTAAACCTATGGAATA[C/T]GGTCTTTCCCTATAAAGGGAGGTCC
Phenotype
MIM: 608214
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GRAMD1B PubMed Links

Gene Details

Gene
GRAMD1B
Gene Name
GRAM domain containing 1B
There are no transcripts associated with this gene.

Gene
SCN3B
Gene Name
sodium voltage-gated channel beta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040151.1 3310 UTR 3 NP_001035241.1
NM_018400.3 3310 UTR 3 NP_060870.1
XM_011542897.1 3310 Intron XP_011541199.1

View Full Product Details