Product Details

SNP ID
rs77714492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124389939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAACATTGGCACAGAGAAAATA[G/T]GAAGACTAATAAAGTGTGGAAAATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8B2 PubMed Links

Gene Details

Gene
OR8B2
Gene Name
olfactory receptor family 8 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005468.1 Intron NP_001005468.1
XM_017017535.1 Intron XP_016873024.1
XM_017017536.1 Intron XP_016873025.1
Gene
OR8B3
Gene Name
olfactory receptor family 8 subfamily B member 3
There are no transcripts associated with this gene.

View Full Product Details