Product Details

SNP ID
rs75620738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:103908877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGAGGGGTTTTGCAAAGAAAA[A/G]CAATTCAAAATTCTGTTACAGCTTA
Phenotype
MIM: 609673
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDGFD PubMed Links

Gene Details

Gene
PDGFD
Gene Name
platelet derived growth factor D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025208.4 2284 UTR 3 NP_079484.1
NM_033135.3 2284 UTR 3 NP_149126.1

View Full Product Details