Product Details

SNP ID
rs74960099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56496461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCACGCATGCCCACAGGCCAC[A/G]GCCTCCACATCACAAACCCCCCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKFZp434L192 PubMed Links

Gene Details

Gene
DKFZp434L192
Gene Name
uncharacterized protein DKFZp434L192
There are no transcripts associated with this gene.

View Full Product Details