Product Details

SNP ID
rs78851873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112877756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTTGTTGCTAATGATACAAAC[A/G]AGAACATACATGTAATCAGAGGTGG
Phenotype
MIM: 125265 MIM: 182175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
REEP5 PubMed Links

Gene Details

Gene
REEP5
Gene Name
receptor accessory protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005669.4 1744 UTR 3 NP_005660.4
XM_017009844.1 1744 Intron XP_016865333.1
Gene
SRP19
Gene Name
signal recognition particle 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204193.1 1744 Intron NP_001191122.1
NM_001204194.1 1744 Intron NP_001191123.1
NM_001204196.1 1744 Intron NP_001191125.1
NM_001204199.1 1744 Intron NP_001191128.1
NM_003135.2 1744 Intron NP_003126.1

View Full Product Details