Product Details

SNP ID
rs76799391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115204654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACCTTGTTGTTGAGTTTATGA[A/C]TGGCTACACTGTTTCTGGATGGCAA
Phenotype
MIM: 602031
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PGGT1B PubMed Links

Gene Details

Gene
PGGT1B
Gene Name
protein geranylgeranyltransferase type I subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005023.3 1235 Intron NP_005014.2
XM_005272020.3 1235 UTR 3 XP_005272077.2
XM_011543490.2 1235 Missense Mutation AGT,ATT S401I XP_011541792.1
XM_011543492.2 1235 Missense Mutation AGT,ATT S278I XP_011541794.1

View Full Product Details