Product Details

SNP ID
rs76647330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:119273154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTGGACAATCTACTTAGCTCT[A/C]TGAGCCTGTTTCTTCACCAGAGTAA
Phenotype
MIM: 612111
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TNFAIP8 PubMed Links

Gene Details

Gene
TNFAIP8
Gene Name
TNF alpha induced protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077654.2 Intron NP_001071122.1
NM_001286813.1 Intron NP_001273742.1
NM_001286814.1 Intron NP_001273743.1
NM_001286815.1 Intron NP_001273744.1
NM_001286817.1 Intron NP_001273746.1
NM_014350.3 Intron NP_055165.2
XM_017009327.1 Intron XP_016864816.1
XM_017009328.1 Intron XP_016864817.1

View Full Product Details