Product Details
- SNP ID
-
rs75246452
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:134741130 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTGAACTCCCTCAGCGTTCCTTC[C/T]GTTTCAAAGCGAGTAGTGCTGGGTG
- Phenotype
-
MIM: 601118
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAMLG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12657663] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMLG
- Gene Name
- calcium modulating ligand
View Full Product Details