Product Details

SNP ID
rs75922891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137753551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGATATCCTCCTTGGGGACTGC[C/T]TTCTTCACCTCCACGCGATGGCCCT
Phenotype
MIM: 609409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA0 PubMed Links

Gene Details

Gene
HNRNPA0
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006805.3 800 Silent Mutation AAA,AAG K172K NP_006796.1

View Full Product Details