Product Details
- SNP ID
-
rs78409055
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:7766741 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTAACTCCCAGCGGAACTGGCA[C/T]GACTCCGTCACCGCCTGCCAGGAAG
- Phenotype
-
MIM: 605872
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC4M
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2277998] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLEC4M
- Gene Name
- C-type lectin domain family 4 member M
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144904.1 |
919 |
Silent Mutation |
CAC,CAT |
H239H |
NP_001138376.1 |
NM_001144905.1 |
919 |
Silent Mutation |
CAC,CAT |
H266H |
NP_001138377.1 |
NM_001144906.1 |
919 |
Silent Mutation |
CAC,CAT |
H154H |
NP_001138378.1 |
NM_001144907.1 |
919 |
Silent Mutation |
CAC,CAT |
H223H |
NP_001138379.1 |
NM_001144908.1 |
919 |
Silent Mutation |
CAC,CAT |
H198H |
NP_001138380.1 |
NM_001144909.1 |
919 |
Silent Mutation |
CAC,CAT |
H244H |
NP_001138381.1 |
NM_001144910.1 |
919 |
Silent Mutation |
CAC,CAT |
H267H |
NP_001138382.1 |
NM_001144911.1 |
919 |
Silent Mutation |
CAC,CAT |
H262H |
NP_001138383.1 |
NM_014257.4 |
919 |
Silent Mutation |
CAC,CAT |
H290H |
NP_055072.3 |
XM_006722611.2 |
919 |
Silent Mutation |
CAC,CAT |
H289H |
XP_006722674.1 |
XM_006722612.3 |
919 |
Silent Mutation |
CAC,CAT |
H262H |
XP_006722675.1 |
XM_006722613.3 |
919 |
Silent Mutation |
CAC,CAT |
H262H |
XP_006722676.1 |
XM_006722614.3 |
919 |
Silent Mutation |
CAC,CAT |
H234H |
XP_006722677.1 |
XM_006722615.2 |
919 |
Silent Mutation |
CAC,CAT |
H290H |
XP_006722678.1 |
View Full Product Details