Product Details
- SNP ID
-
rs74959615
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8056212 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCATTGGGTGGGCTGTGCTCCGGC[A/G]CGCCTGGACTTACCGGATCCAGGAG
- Phenotype
-
MIM: 602565
MIM: 608529
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCL25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143413416] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCL25
- Gene Name
- C-C motif chemokine ligand 25
- Gene
- FBN3
- Gene Name
- fibrillin 3
There are no transcripts associated with this gene.
View Full Product Details