Product Details

SNP ID
rs79711434
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:145892356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTTTCAAGCCAACGTCAGCTC[T/C]CTCCTGTGGAGTGGGGAACAGGCAG
Phenotype
MIM: 604042
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ANKRD35 PubMed Links

Gene Details

Gene
ANKRD35
Gene Name
ankyrin repeat domain 35
There are no transcripts associated with this gene.

Gene
ITGA10
Gene Name
integrin subunit alpha 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303040.1 3528 UTR 3 NP_001289969.1
NM_001303041.1 3528 UTR 3 NP_001289970.1
NM_003637.4 3528 UTR 3 NP_003628.2
XM_005277436.3 3528 UTR 3 XP_005277493.1
XM_011510083.2 3528 UTR 3 XP_011508385.1
XM_011510084.1 3528 UTR 3 XP_011508386.1
XM_017002622.1 3528 UTR 3 XP_016858111.1
XM_017002623.1 3528 UTR 3 XP_016858112.1
XM_017002624.1 3528 UTR 3 XP_016858113.1
XM_017002625.1 3528 UTR 3 XP_016858114.1
XM_017002626.1 3528 UTR 3 XP_016858115.1
XM_017002627.1 3528 Intron XP_016858116.1
XM_017002628.1 3528 UTR 3 XP_016858117.1

View Full Product Details