Product Details

SNP ID
rs77641164
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:220095511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTTTTAGTCAATACCTTCTGT[A/G]TGCCAGAAAGAGTGACAGTTTCAAA
Phenotype
MIM: 604053 MIM: 612801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPNT1 PubMed Links

Gene Details

Gene
BPNT1
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
There are no transcripts associated with this gene.

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018060.3 Intron NP_060530.3

View Full Product Details