Product Details
- SNP ID
-
rs75277931
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:224388132 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTGTTTTTCTTTCTTCTCCCCTT[C/T]CCTGACTTTTCCCTTTCTCCCATCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNIH4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5781364] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNIH4
- Gene Name
- cornichon family AMPA receptor auxiliary protein 4
There are no transcripts associated with this gene.
- Gene
- WDR26
- Gene Name
- WD repeat domain 26
View Full Product Details