Product Details

SNP ID
rs80299691
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:225404482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGAACAAAGCCCCACCATCCAG[A/C]AACTAGAAGATTTTTTCCCGTTGAA
Phenotype
MIM: 603341 MIM: 600024
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
There are no transcripts associated with this gene.

Gene
LBR
Gene Name
lamin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002296.3 1613 Missense Mutation GCT,TCT A537S NP_002287.2
NM_194442.2 1613 Missense Mutation GCT,TCT A537S NP_919424.1
XM_005273125.2 1613 Missense Mutation GCT,TCT A495S XP_005273182.1
XM_011544185.2 1613 Missense Mutation GCT,TCT A537S XP_011542487.1

View Full Product Details