Product Details

SNP ID
rs78656147
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:214351670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATCAGCAAATCCAGGCAAATAT[G/T]ATAAACCACACATCATAATCCATGA
Phenotype
MIM: 603155
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTPN14 PubMed Links

Gene Details

Gene
PTPN14
Gene Name
protein tyrosine phosphatase, non-receptor type 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005401.4 10469 UTR 3 NP_005392.2
XM_017001941.1 10469 Intron XP_016857430.1

View Full Product Details