Product Details

SNP ID
rs77824904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30103220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTTAAAAAGAATGAGGAAGAG[C/T]TGTAAGTGCTAACATGTAAATATGT
Phenotype
MIM: 607338
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LEPROTL1 PubMed Links

Gene Details

Gene
LEPROTL1
Gene Name
leptin receptor overlapping transcript-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128208.1 Intron NP_001121680.1
NM_015344.2 Intron NP_056159.2

View Full Product Details